Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen peroxide solution, Solstice

Catalog No. 60090331 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-090-331 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-331 Supplier Solstice Supplier No. 40267H5L

Semiconductor grade ( 17948), ≥30%

  • comprehensive portfolio for the electronics and semiconductor industry
  • for scientists engaged in education, research, development, prototyping and small scale fabrication of high tech micro- and nanochips
  • Superior purity, stability and exceptional lot-to-lot consistency
  • includes high purity products required for high performance applications such as etchants, solvents or hexamethyldisilazane (HMDS)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-84-1 , 7732-18-5
Molecular Formula H2O2
Molecular Weight (g/mol) 34.01
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name peroxol
SMILES OO

Specifications

CAS Min % 68.90%, 30.00%
Boiling Point Decomposes on heating.
CAS Max % 69.90%, 31.00%
Packaging Plastic Bottle
Quantity 5 L
Physical Form Liquid
Grade Semiconductor grade reagent
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.