Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrogen Potassium ATPase Beta Rabbit anti-Human, Mouse, Rat, Clone: 3Y1Y1, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP31573920UL
This item is not returnable.
View return policy
Description
Hydrogen Potassium ATPase Beta Monoclonal antibody specifically detects Hydrogen Potassium ATPase Beta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Hydrogen Potassium ATPase Beta | |
Monoclonal | |
Unconjugated | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
3Y1Y1 | |
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
ATP6B, ATPase, H+/K+ exchanging, beta polypeptide, ATPase, H+/K+ transporting, beta polypeptide, Gastric H(+)/K(+) ATPase subunit beta, gastric H+/K+ ATPase beta subunit, gastric hydrogen-potassium ATPase, beta, potassium-transporting ATPase beta chain, potassium-transporting ATPase subunit beta, Proton pump beta chain | |
A synthetic peptide corresponding to a sequence within amino acids 192-291 of human Hydrogen Potassium ATPase Beta (P51164). | |
20 μg | |
Endocrinology, Signal Transduction | |
496 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction