Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen tetrachloroaurate hydrate, 99.999%, (trace metal basis)

Catalog No. p-4228713
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 27988-77-8 | AuCl4H | 339.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27988-77-8
Molecular Formula AuCl4H
Molecular Weight (g/mol) 339.77
MDL Number MFCD00149903
InChI Key RFWCEGCWRSNAGT-UHFFFAOYSA-K
Synonym chloroauric acid hydrate, Gold trichloride acid hydrate
IUPAC Name hydrogen tetrachlorogoldtris(ylium)
SMILES [H+].Cl[Au+3](Cl)(Cl)Cl

Specifications

Melting Point 30.0°C
Color Yellow to Orange
Physical Form Crystals
Quantity 1 g
Assay Percent Range 49 to 51% (Au)
Linear Formula HAuCl4·xH2O
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 339.78
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material hydrogen tetrachloroaurate hydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.