Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen tetrachloroaurate(III) trihydrate, ACS, 99.99% (metals basis), Au 49.0% min

Catalog No. AA3640006
Click to view available options
Quantity:
1 g
5 g

CAS: 16961-25-4 | AuCl4H | 339.77 g/mol

It is used for microanalysis of rubidium, cesium and in the determination of alkaloids. Usually used as a harbinger for the preparation of gold (Au) nanoparticles. It is used in the control rods of nuclear reactors and in the manufacturing of electric light filaments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16961-25-4
Molecular Formula AuCl4H
Molecular Weight (g/mol) 339.77
MDL Number MFCD00149904
InChI Key VDLSFRRYNGEBEJ-UHFFFAOYSA-K
Synonym Chloroauric acid
IUPAC Name trichlorogold hydrochloride
SMILES Cl.Cl[Au](Cl)Cl

Specifications

Physical Form Crystalline
Quantity 5 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in water. Insoluble in ether.
Formula Weight 393.83 (339.79 Anhydrous)
Grade ACS Reagent
Sensitivity Hygroscopic; Light sensitive
Chemical Name or Material Hydrogen tetrachloroaurate trihydrate, For analysis ACS
Hazard Category H302-H314-H317-H335
Precautionary Statement P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 240-948-4
RTECSNumber MD5428000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.