Learn More
United Scientific™ Hydrometer
SDPSCI_ED
Ideal for classroom use.
$6.95 - $7.10
Specifications
Type Hydrometer
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Application) Price Quantity & Availability  
S15897
SDPSCI_ED
View Documents
United Scientific™
HYMH02
Heavy Liquids
Each for $7.10
Only null left
Add to Cart
S15917
SDPSCI_ED
View Documents
United Scientific™
HYML01
Light Liquids
Each for $6.95
Only null left
Add to Cart
Description
Description
  • Hydrometer, used to determine the specific gravity of liquids. Two specific gravity ranges are available.
  • HYMH02 is for use with heavy liquids
  • HYML01 is for use with light liquids
  • Each is housed in a reusable storage tube
Specifications
Specifications
Hydrometer
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.