Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Hydromorphone Polyclonal Antibody
Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA175167
Description
Working Concentration: 1.25 μg/mL. Sensitivity: 10 ng/mL Hydromorphone produces 74% inhibition in a competitive ELISA employing Hydromorphone polyclonal antibody.
Hydromorphone Base is the hydrogenated ketone of morphine, a semi-synthetic opioid with analgesic effects. Hydromorphone selectively binds the mu-opioid receptor which is linked through G-proteins. Binding stimulates the exchange of guanosine triphosphate (GTP) for guanosine diphosphate (GDP) on the G-protein complex and interacts with and inhibits adenylate cyclase located at the inner surface of the plasma membrane. This leads to a reduction in intracellular cyclic 3',5'-adenosine monophosphate (cAMP). Further, voltage-gated potassium channels are activated, thereby causing hyperpolarization and reducing neuronal excitability. In addition, the opening of voltage-gated calcium channels is inhibited, thereby leading to an inhibition of calcium entry and a reduction in the release of various neurotransmitters, including GABA, vasopressin, somatostatin, insulin and glucagons.
Specifications
Hydromorphone | |
Polyclonal | |
Unconjugated | |
C17H19NO3; Dihydromorfinon; Dihydromorphinone; Dilaudid; Dimorphone; Idromorfone | |
Hydromorphone(3)-BTG. | |
1 mg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA | |
7.28 mg/mL | |
PBS with 0.09% sodium azide; pH 7.8 | |
Sheep | |
purified | |
RUO | |
Chemical | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction