Learn More
Hydroquinone, 99%
Description
Hydroquinone is used as a reducing agent and finds applications in black and white photographic developers for film and paper along with 4-(methylamino)phenol sulfate (metol). In this application, silver halides are reduced to elemental silver. It acts as a polymerization inhibitor which prevents polymerization of acrylic acid, methyl methacrylate, cyanoacrylate and other monomers. It undergoes mild oxidation to convert into parabenzoquinone. It is also used as an intermediate to produce antioxidants for rubber and food.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Hydroquinone is used as a reducing agent and finds applications in black and white photographic developers for film and paper along with 4-(methylamino)phenol sulfate (metol). In this application, silver halides are reduced to elemental silver. It acts as a polymerization inhibitor which prevents polymerization of acrylic acid, methyl methacrylate, cyanoacrylate and other monomers. It undergoes mild oxidation to convert into parabenzoquinone. It is also used as an intermediate to produce antioxidants for rubber and food.
Solubility
Soluble in water, methanol, ethanol, diethyl ether, acetone, benzene and carbon tetrachloride.
Notes
Air and light sensitive. Incompatible with strong bases and strong oxidizing agents.

Specifications
Specifications
Melting Point | 170°C to 172°C |
Density | 1.32 |
pH | 3.75 |
Boiling Point | 285°C to 287°C |
Flash Point | 165°C (329°F) |
Odor | Odorless |
Quantity | 5000 g |
UN Number | UN3077 |
Beilstein | 605970 |
Sensitivity | Air and light sensitive |
Show More |
Safety and Handling
Hazard Category | H302-H317-H318-H335-H341-H351-H500 |
Hazard Statement | GHS H Statement H318-H341-H351-H302-H317 Causes serious eye damage. Suspected of causing genetic defects. Suspected of causing cancer. Harmful if swallowed. May cause an allergic skin reaction. |
Precautionary Statement | P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P363-P501c |
DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
EINECSNumber | 204-617-8 |
RTECSNumber | MX3500000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.