Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroquinone, 99%

Catalog No. AC120910100
Click to view available options
Quantity:
500 g
2 kg
10 kg
Packaging:
Plastic bottle
Plastic drum

CAS: 123-31-9 | C6H6O2 | 110.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-31-9
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.11
MDL Number MFCD00002339
InChI Key QIGBRXMKCJKVMJ-UHFFFAOYSA-N
Synonym hydroquinone, 1,4-benzenediol, quinol, 1,4-dihydroxybenzene, p-benzenediol, 4-hydroxyphenol, p-hydroquinone, p-hydroxyphenol, p-dihydroxybenzene, benzoquinol
PubChem CID 785
ChEBI CHEBI:17594
IUPAC Name benzene-1,4-diol
SMILES C1=CC(=CC=C1O)O

Specifications

Melting Point 170.0°C to 174.0°C
Color White
Density 1.32
Boiling Point 285.0°C to 287.0°C
Flash Point 165°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic drum
Quantity 10 kg
Beilstein 06, 836
Fieser 05,341; 14,249
Merck Index 15, 4845
Solubility Information Solubility in water: 70g/L in water (20°C). Other solubilities: soluble in alcohol and ether,slightly soluble in benzene,readily soluble in ethanol,acetone and methanol
Formula Weight 110.11
Percent Purity 99%
Physical Form Needle-like Crystals or Crystalline Powder
Chemical Name or Material Hydroquinone, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Suspected of causing cancer.
Harmful if swallowed.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-617-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.