Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydroquinone bis(2-hydroxyethyl) ether, 95%
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
Hydroquinone bis(2-hydroxyethyl) ether is used to produce 1,4-bis-(2-bromo-ethoxy)-benzene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Hydroquinone bis(2-hydroxyethyl) ether is used to produce 1,4-bis-(2-bromo-ethoxy)-benzene.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. Protect from humidity and water. It is hygroscopic in nature. Incompatible with oxidizing agents.
Hydroquinone bis(2-hydroxyethyl) ether is used to produce 1,4-bis-(2-bromo-ethoxy)-benzene.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. Protect from humidity and water. It is hygroscopic in nature. Incompatible with oxidizing agents.

Specifications
Specifications
Melting Point | 100°C to 102°C |
Quantity | 2500 g |
Sensitivity | Hygroscopic |
Solubility Information | Insoluble in water. |
Formula Weight | 198.22 |
Percent Purity | 95% |
Chemical Name or Material | Hydroquinone bis(2-hydroxyethyl) ether |
Safety and Handling
EINECSNumber | 203-197-3 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction