Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HydroSpec™ Solvent, Ricca Chemical

Click to view available options
Quantity:
1 L
Specifications
Specifications
Chemical Name or Material | Solvent |
CAS Min % | 5, 10 |
CAS Max % | 10, 95 |
CAS | 7446-09-5, 288-32-4, 67-56-1 |
Physical Form | Liquid |
Quantity | 1 L |
Packaging | Amber Glass Bottle |
UN Number | UN1230 |
Specific Gravity | 0.91 |
Grade | Karl Fischer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction