Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HydroSpec™ Titrant T2, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
1 L
6 x 1 L
Specifications
Specifications
CAS Min % | 85, 1 |
CAS | 67-56-1, 7553-56-2 |
Physical Form | Liquid |
Quantity | 1 L |
CAS Max % | 5 |
Packaging | Amber Glass Bottle |
UN Number | UN1230 |
Specific Gravity | 0.89 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction