Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydroxyacid Oxidase-1/HAO-1 Antibody (001), DyLight 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290046DL594
Description
Hydroxyacid Oxidase-1/HAO-1 Monoclonal specifically detects Hydroxyacid Oxidase-1/HAO-1 in Human samples. It is validated for ELISA.Specifications
Hydroxyacid Oxidase-1/HAO-1 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
1 | |
ELISA | |
(S)-2-hydroxy-acid oxidase, EC 1.1.3.15, Glycolate oxidase, GOX1MGC142227, GOXMGC142225, HAOX1, hydroxyacid oxidase (glycolate oxidase) 1, hydroxyacid oxidase 1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Hydroxyacid Oxidase-1/HAO-1 (NP_060015.1; Leu 2-Ile 370). | |
0.1 mL | |
metabolism, Signal Transduction | |
54363 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction