Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydroxyacid Oxidase-1/HAO-1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299929MFV450
Description
Hydroxyacid Oxidase-1/HAO-1 Polyclonal antibody specifically detects Hydroxyacid Oxidase-1/HAO-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)Specifications
Hydroxyacid Oxidase-1/HAO-1 | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry-Paraffin | |
(S)-2-hydroxy-acid oxidase, EC 1.1.3.15, Glycolate oxidase, GOX1MGC142227, GOXMGC142225, HAOX1, hydroxyacid oxidase (glycolate oxidase) 1, hydroxyacid oxidase 1 | |
Produced in rabbits immunized with purified, recombinant Human Hydroxyacid Oxidase-1/HAO-1 (Accession#: NP_060015.1; Leu2-Ile370) | |
0.1 mL | |
metabolism, Signal Transduction | |
54363 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction