Learn More
Description
Autophagy inhibitor. Inhibits growth and induces apoptosis of renal cancer cells in vitro. Also inhibits TLR9. Additionally inhibits PRC2 by inhibiting the allosteric binding of PRC2 to EED (embryonic ectoderm development) within the H3K27me3-binding pocket, and reduces H3K27me3 levels in multiple myeloma cells in vitro.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Chemical Name or Material | 2-[[4-[(7-Chloro-4-quinolinyl)amino]pentyl]ethylamino]ethanol sulfate |
| CAS | 747-36-4 |
| Quantity | 10 mg |
| Target | Autophagy-related Compounds |
| Molecular Formula | C18H28ClN3O5S |
| Formula Weight | 433.95 |
| Purity | >99% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.


