Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxychloroquine Sulfate 98.0+%, TCI America™
SDP

Catalog No. H130625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H130625G 25 g
H13065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H130625G Supplier TCI America Supplier No. H130625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 747-36-4
Molecular Formula C18H28ClN3O5S
Molecular Weight (g/mol) 433.948
MDL Number MFCD00078203
InChI Key JCBIVZZPXRZKTI-UHFFFAOYSA-N
Synonym hydroxychloroquine sulfate, hydroxychloroquine sulphate, ercoquin, plaquinol, toremonil, 2-4-7-chloroquinolin-4-yl amino pentyl ethyl amino ethanol sulfate, hydroxychloroquine sulfate usp, plaquenil tn, hydroxychloroquine; sulfuric acid, dsstox_cid_27788
PubChem CID 12947
IUPAC Name 2-[4-[(7-chloroquinolin-4-yl)amino]pentyl-ethylamino]ethanol;sulfuric acid
SMILES CCN(CCCC(C)NC1=C2C=CC(=CC2=NC=C1)Cl)CCO.OS(=O)(=O)O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 433.95
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Hydroxychloroquine Sulfate
RTECSNumber KK3342500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.