Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxyethyl Cellulose, 3,400 cps, Spectrum™ Chemical
SDP

Catalog No. 18603307
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

H1148, 9004-62-0

Spectrum™ Chemical Hydroxyethyl Cellulose, 3,400 cps is a thickening and gelling agent created from cellulose, most commonly used in cleaning solutions, cosmetics and other products used in the household. It usually comes in capsule form. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 9004-62-0
Molecular Formula C29H52O21
Molecular Weight (g/mol) 736.71
InChI Key CWSZBVAUYPTXTG-UHFFFAOYNA-N
IUPAC Name 5-{[6-({[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}methyl)-3,4-dihydroxy-5-{[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}oxan-2-yl]oxy}-6-(hydroxymethyl)-2-methyloxane-3,4-diol
SMILES COC1C(O)C(O)C(OCC2OC(OC3C(O)C(O)C(C)OC3CO)C(O)C(O)C2OC2OC(CO)C(OC)C(O)C2OCCO)OC1CO

Specifications

CAS Max % 100%
Loss on Drying 5%
Packaging Fiber Drum
Quantity 45 kg
Grade Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.