Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydroxylamine Hydrochloride, 10% (w/w), Ricca Chemical

Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
Specifications
Specifications
Color | Colorless |
CAS Min % | 88.0, 9.81 |
CAS Max % | 92.0, 10.21 |
Percent Purity | 10% |
Grade | Laboratory |
Packaging | Natural Poly Bottle |
Solvent or Matrix | Aqueous Solution |
pH | 3.2 |
Quantity | 1 L |
Physical Form | Liquid |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction