Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxylamine sulfate, 99%

Catalog No. p-3736592
Click to view available options
Quantity:
100 g
1 kg
5 kg
Packaging:
Plastic bottle

CAS: 10039-54-0 | H6N2O2·H2SO4 | 164.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10039-54-0
Molecular Formula H6N2O2·H2SO4
Molecular Weight (g/mol) 164.14
MDL Number MFCD00044869
Synonym Hydroxylammonium sulfate

Specifications

Melting Point 170.0°C
Color White
Physical Form Crystals
Quantity 100 g
Assay Percent Range 99%
Linear Formula (H2NOH)2·H2SO4
Fieser 01,481
Merck Index 15, 4867
Solubility Information Solubility in water: 329g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 164.14
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Plastic bottle
Chemical Name or Material Hydroxylamine sulfate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May be corrosive to metals.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of causing ca
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persists: Get medi
EINECSNumber 233-118-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.