Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxylapatite, MP Biomedicals™

Catalog No. p-4404639 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
10 g
25 g
50 g
100 g

Chemical Identifiers

CAS 1306-06-5
Molecular Formula Ca5HO13P3
Molecular Weight (g/mol) 502.31
MDL Number MFCD00010904
InChI Key XYJRXVWERLGGKC-UHFFFAOYSA-D
Synonym Calcium phosphate hydroxide, Durapatite
IUPAC Name pentacalcium hydroxide triphosphate
SMILES [OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 10 g
Formula Weight 502.31
Chemical Name or Material Hydroxylapatite
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.