Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxyprogesterone Caproate, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog No. 18603330 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
5 kg
Packaging:
Poly Pail

H1234, 630-56-8, C27H40O4

Spectrum™ Chemical Hydroxyprogesterone Caproate, USP is a prescription hormone medication used to lower the risk of preterm birth in women who are pregnant with one baby and who have delivered one baby too early (preterm) in the past. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

CAS 630-56-8
CAS Max % 100%
Melting Range 120°-124°C
Packaging Poly Pail
Molecular Formula C27H40O4
Quantity 5 kg
Specific Rotation +58° to +64°
InChI Key DOMWKUIIPQCAJU-LJHIYBGHSA-N
SMILES CCCCCC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)=O
IUPAC Name (1R,3aS,3bR,9aR,9bS,11aS)-1-acetyl-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl hexanoate
Molecular Weight (g/mol) 428.61
Percent Purity 97 to 103%
Grade USP
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.