Learn More
Hydroxypropyl methacrylate, mixture of isomers, 97+%, stab. with ca 0.02% 4-methoxyphenol
Description
Hydroxypropyl methacrylate is a functional monomer for the preparation of hot solid acrylic coatings, styrene-butadiene latex modifiers, acrylic modified polyurethane coating ,water-soluble plating coatings , adhesives ,textile finishing agent ,paper coating , photosensitive paint and polyurethane vinyl resin modified agent. Mainly used in the manufacture are active groups of hydroxyl acrylic resin. Used in reactive diluent and cross-linking agent in the UV curing system.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Hydroxypropyl methacrylate is a functional monomer for the preparation of hot solid acrylic coatings, styrene-butadiene latex modifiers, acrylic modified polyurethane coating ,water-soluble plating coatings , adhesives ,textile finishing agent ,paper coating , photosensitive paint and polyurethane vinyl resin modified agent. Mainly used in the manufacture are active groups of hydroxyl acrylic resin. Used in reactive diluent and cross-linking agent in the UV curing system.
Solubility
Soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 70°C (1mmHg) |
| Density | 1.029 |
| Boiling Point | 205°C to 209°C |
| Flash Point | 97°C (207°F) |
| Refractive Index | 1.448 |
| Quantity | 5000 mL |
| Sensitivity | Light sensitive |
| Solubility Information | Soluble in water. |
| Formula Weight | 144.17 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | H317-H319 |
| Hazard Statement | GHS H Statement H319-H317 Causes serious eye irritation. May cause an allergic skin reaction. |
| Precautionary Statement | P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P363-P501c |
| EINECSNumber | 248-666-3 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.