Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hydroxysteroid (17-beta) dehydrogenase 11 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299361MFV450
Description
hydroxysteroid (17-beta) dehydrogenase 11 Polyclonal antibody specifically detects hydroxysteroid (17-beta) dehydrogenase 11 in Human samples. It is validated for Western BlotSpecifications
hydroxysteroid (17-beta) dehydrogenase 11 | |
Polyclonal | |
Western Blot | |
17betaHSD11, 17betaHSDXI, 17-beta-hydroxysteroid dehydrogenase 11, 17-beta-hydroxysteroid dehydrogenase type XI, 17-beta-hydroxysteroid dehydrogenase XI, 17bHSD11, CTCL-associated antigen HD-CL-03, dehydrogenase/reductase (SDR family) member 8, DHRS8, EC 1.1.1, EC 1.1.1.62, hydroxysteroid (17-beta) dehydrogenase 11, member 2,17-beta-HSD 11, PAN1BRETSDR2,17-BETA-HSD11, retSDR2, RetSDR2,17-BETA-HSDXI, SDR16C2,17BHSD11, T-cell lymphoma-associated antigen HD-CL-03,17-beta-HSD XI | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human hydroxysteroid (17-beta) dehydrogenase 11. | |
0.1 mL | |
Lipid and Metabolism | |
51170 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction