Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxyzine Hydrochloride, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18603367
Click to view available options
Quantity:
100 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum

HY125, 2192-20-3, C21H27ClN2O2·2HCl

Spectrum™ Chemical Hydroxyzine Hydrochloride, USP is an antihistamine and has strong anxiolytic and antipsychotic properties. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 2192-20-3
Molecular Formula C21H29Cl3N2O2
Molecular Weight (g/mol) 447.83
InChI Key ANOMHKZSQFYSBR-UHFFFAOYNA-N
IUPAC Name 2-(2-{4-[(4-chlorophenyl)(phenyl)methyl]piperazin-1-yl}ethoxy)ethan-1-ol dihydrochloride
SMILES Cl.Cl.OCCOCCN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1

Specifications

CAS Max % 100%
Ignition Residue 0.5%
Packaging Fiber Drum
Quantity 25 kg
Percent Purity 98 to 102%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.