Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ HyFlex™ 11-280 Sleeves
GREENER_CHOICE

Catalog No. 19500733 Shop All Ansell Products
Change view
Click to view available options
Size:
Narrow
Wide
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-500-733 Narrow
19-500-731 Narrow
19-500-732 Wide
19-500-734 Wide
19-500-735 Narrow
19-500-736 Wide
19-500-737 Narrow
19-500-738 Wide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-500-733 Supplier Ansell™ Supplier No. 11280160N
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ansell™ HyFlex™ 11-280 Sleeves feature a seamless design and options of lengths and widths to ensure an excellent fit for all-day comfort

  • INTERCEPT™ Cut Resistance Technology provides best-in-class cut protection and a soft and cool feel to the worker
  • Engineered to perform in combination with any HyFlex cut resistant glove
  • Available in three versions: Extra Wide, Wide, Narrow
TRUSTED_SUSTAINABILITY

Specifications

Product Type Sleeve
Size Narrow
Construction Knitted
Material HPPE Yarn
Color Gray
Sterility Non-sterile
Style Ambidextrous
Disposable Reusable
Closure Type Elastic Upper with Knit Cuff
Seam Type Seamless
Latex Free Yes
Model HyFlex™11-280
For Use With (Application) Glass repair and replacement; Handling metal sheets and panels
Certifications/Compliance REACH Compliant
Certifications/Compliance (Europe) EN420:2003 + A1:2009; Category ll
Length (Metric) 40 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.