Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ HyFlex™ 11-281 Sleeves
GREENER_CHOICE

Catalog No. 19500744 Shop All Ansell Products
Change view
Click to view available options
Size:
Narrow
Wide
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-500-744 Wide
19-500-741 Narrow
19-500-742 Wide
19-500-743 Narrow
19-500-745 Narrow
19-500-746 Wide
19-500-747 Narrow
19-500-748 Wide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-500-744 Supplier Ansell™ Supplier No. 11281160W
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ansell™ HyFlex™ 11-281 Sleeves feature a seamless design and options of lengths and widths to ensure an excellent fit for all-day comfort

  • INTERCEPT™ Cut Resistance Technology provides best-in-class cut protection and a soft and cool feel to the worker
  • Engineered to perform in combination with any HyFlex cut resistant glove
  • Available in three versions: Extra Wide, Wide, Narrow
TRUSTED_SUSTAINABILITY

Specifications

Product Type Sleeve
Size Wide
Construction Knitted
Material HPPE Yarn
Color Gray
Sterility Non-sterile
Disposable Reusable
Model HyFlex™11-281
For Use With (Application) Glass repair and replacement; Handling metal sheets and panels
Certifications/Compliance REACH Compliant
Certifications/Compliance (Europe) EN420:2003 + A1:2009; Category ll
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.