Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell HyFlex™ 11-435 Cut-Resistant Glove, Palm Coated Polyurethane and Nitrile

Catalog No. 19010217 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-010-217 8
19-010-215 6
19-010-216 7
19-010-218 9
19-010-219 10
19-010-220 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-010-217 Supplier Ansell Supplier No. 288906
This item is not returnable. View return policy

Knit gloves with palm-coated finishing provide extreme resistance to cuts and burrs while providing a comfortable fit, flexibility and cool feeling.

  • Unique combination of Dyneema, Nylon, Lycra, and glass fiber
  • ISO Cut level C
  • Water-based polyurethane and NBR
  • Ultra strong fibers provide extreme resistance to cuts and burrs
  • Anti-static for improved safety in critical areas
  • Silicone free
  • Oeko-Tex 100 approved
  • Also available in vend pack
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 5
Certifications/Compliance (Europe) CE, EN 420:2003 + A1:2009; Category ll
Construction Knit
Coating Material Nitrile
Color Black, Gray
Cut Resistance Rating ANSI A3
Hand Specific
Latex Free Yes
Product Type Cut-resistant Gloves
Size 8
EN388 Standard EN 388:4542; EN 388:4X42C
For Use With (Application) Handling metal sheets and panels, handling sharp and rough edges, body painting, mounting and dismantling, foundry operations, maintenance
Glove Cuff Style Knit
Liner Material Nylon, Lycra™, Fiberglass, Dyneema™
Model 288906
Quantity 12 Pairs/Bag, 12 Bags/Cs.
Series HyFlex™ 11-435
Length (English) 8.8 in.
Length (Metric) 22.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.