Learn More
Ansell Hyflex™ 11-561 Vend Pack Gloves
Catalog No. 17004257 Shop All Ansell Products
Change view
Click to view available options
Size:
5
6
7
8
9
10
11
12
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-004-257 9
17-004-253 5
17-004-254 6
17-004-255 7
17-004-256 8
17-004-258 10
17-004-259 11
17-004-260 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 17-004-257 Supplier Ansell Supplier No. 11561VP090
Add to Cart
Add to Cart
This item is not returnable. View return policy
Our lightest EN ISO Cut C and ANSI/ISEA 105-2024 Cut A3 glove. Offering 2 times greater cut resistance.
  • Ultra-lightweight design: 20% lighter for unparalleled comfort and dexterity
  • EN CUT ISO C and ANSI/ ISEA 105-2024 CUT A3 protection: 2x greater cut resistance
  • Improved FORTIX™ Technology: up to 20% greater durability
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 388:2016, Dermatologically Tested, Ecovadis label, EN 388:2016 +A1:2018, EN ISO 21420:2020, Food Contact, OEKO-TEX™ Centexbel, Category II
Construction Knit
Cuff Material Nylon, HPPE, Basalt, Spandex, Polyester
Coating Material Nitrile
Color Blue
Cut Resistance Rating ANSI A3
Hand Specific
Latex Free No
Material Nylon, HPPE, Spandex, Polyester, Nitrile
Product Type Cut-resistant Gloves
Size 9
EN388 Standard 4 x 42 C
For Use With (Application) Handling metal sheets and panels, Handling parts with sharp, rough edges, Final assembly, Handling materials with sharp or rough edges, Assembly and inspection of components, Applying finish to materials, products, Compatible with consumer and industrial touchscreens
Gender Unisex
Glove Cuff Color Blue
Glove Cuff Style Knit
Liner Material Nylon, HPPE, Basalt, Spandex, Polyester
Model 11-561
Puncture Rating ANSI/ISEA 105-16 Level 4
Thumb Style Reinforced
Yarn Gauge 15