Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell HyFlex™ 11-618 Extra Thin Polyurethane Coated Nylon Gloves
GREENER_CHOICE

Catalog No. 19010205 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-010-205 8
19-010-203 6
19-010-204 7
19-010-206 9
19-010-207 10
19-010-208 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-010-205 Supplier Ansell Supplier No. 288599
Add to Cart
Add to Cart
This item is not returnable. View return policy

Light duty, 18-gauge gloves offer tactility and dexterity with a 0.8 mm polyurethane-coating for an improved grip and additional abrasion resistance.

  • Extremely thin work gloves offer comfort, fit, and sensitivity for the most delicate handling
  • Designed for a wide range of lightweight applications in dry or slightly oily environments
  • Polyurethane coating provides dexterity and abrasion resistance
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Coating Material Polyurethane
Color Blue
Cut Resistance Rating ANSI A1
Hand Specific
Latex Free No
Material Nylon
Product Type Dip-coated Gloves
Size 8
For Use With (Application) Automotive Assembly, Machinery/Equipment Operation, and Metal Fabrication
Glove Cuff Style Knit
Series 11-618
Yarn Gauge 18
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.