Description
- Features INTERCEPT Technology
- Ultra-thin breathable gloves that offer high cut resistance, while maintaining enhanced flexibility and tactility
- Touchscreen compatible
- Designed to fit the natural shape of the hand to help reduce hand fatigue
- Clean and skin friendly. Dermatologically approved by Dermatest
Specifications
Specifications
| Coating Material | Polyurethane |
| Color | Gray |
| Cut Resistance Rating | ANSI A5 |
| Material | HPPE, Tungsten, Spandex, Polyester |
| Size | 10 |
| Style | HyFlex™ 11-755 |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.