Learn More
Ansell HyFlex™ 11-757 Cut-Resistant Touchscreen Gloves
Catalog No. 17004278 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-004-278 9
17-004-275 6
17-004-276 7
17-004-277 8
17-004-279 10
17-004-280 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-004-278 Supplier Ansell Supplier No. 11757090VP
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ultralight ANSI A7 / ISO F-level cut-resistant touchscreen glove.
  • Extreme cut protection featuring INTERCEPT™ Technology while maintaining enhanced flexibility and tactility
  • Touchscreen compatible to ensure that workers do not need to change or remove gloves between tasks
  • High dexterity combined with unmatched comfort and precision while achieving excellent abrasion resistance
  • Tailored fit at the base of the pinky finger better matches natural hand contours
  • Dermatologically approved by Dermatest™
TRUSTED_SUSTAINABILITY
Specifications
Coating Material Polyurethane
Color Black
Cut Resistance Rating ANSI A7
Latex Free Yes
Material Nylon, Stainless Steel Fiber, Polyester, Tungsten
Size 9
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.