Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell HyFlex™ 11-801 Foam Nitrile Coated Nylon Gloves

Catalog No. 19160411C Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-160-411C 8
19-160-411A 6
19-160-411B 7
19-160-411D 9
19-160-411E 10
19-160-411F 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-160-411C Supplier Ansell Supplier No. 205674
Add to Cart
Add to Cart
This item is not returnable. View return policy

Foam nitrile coated gloves offer an improved design that is up to 30% more breathable to enhance user comfort. The dark colored liner is suitable for slightly dirty or oily environments.

  • Dark colored liner is well suited for slightly dirty or oily environments
  • Foam nitrile coating is up to 30% more breathable for increased user comfort
  • Proprietary washing process efficiently extracts impurities to deliver a cleaner, more comfortable glove
  • Independently certified as skin friendly by the Oeko-Tex™ testing & certification system
  • No transfer of silicone contaminants to metal parts prior to painting
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Coated Area Palm
Coating Material Nitrile Foam
Color Gray
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Nylon
Product Type Dip-coated Gloves
Size 8
For Use With (Application) Automotive, Machinery/Equipment, Metal Fabrication, and Chemical Handling
Glove Cuff Style Knit
Series 11-801
Yarn Gauge 15
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.