Learn More
Ansell HyFlex™ 11581 Light Duty Touchscreen Compatible Cut-resistant Gloves
Click to view available options
Size:
6
7
8
9
10
11
12
Description
- High cut protection with an immediate blade blunting effect outperforming cut protection of gloves in the same category
- High dexterity and tactility
- 21-gauge construction makes it the thinnest and lightest glove in its category
- Touchscreen compatible, so it's ready to use in today's workplaces
- Extended Use-FORTIX™ technology provides up to 20% more abrasion resistance for longer lasting handling in abrasive conditions
- Reinforced thumb crotch
- Made of tungsten, HPPE and nylon
- Spandex seamless liner
- DGX nitrile palm coating
Specifications
Specifications
Coating Material | Nitrile |
Color | Gray, Black |
Cut Resistance Rating | ANSI A6 |
Hand | Specific |
Latex Free | Yes |
Material | Nylon |
Product Type | Cut-resistant Gloves |
Size | 10 |