Learn More
Ansell HyFlex™ 11591 Medium Duty Touchscreen Compatible Cut-resistant Gloves
Shop All Ansell ProductsDescription
- Works with most touchscreen devices
- Comfortable, breathable, seamless knitted liner
- Latex-, silicone-, DMF- and fiber glass-free
- Reinforced thumb crotch
- Improved FORTIX™ Technology delivers up to 20% more abrasion resistance for longer-lasting wear
- 13 Gauge
- Made of HPPE, Nylon, Spandex, Steel, Graphene and Tungsten
Specifications
Specifications
| Coating Material | Nitrile |
| Color | Black |
| Cut Resistance Rating | ANSI A9 |
| Hand | Specific |
| Latex Free | Yes |
| Material | Nylon |
| Product Type | Cut- and Abrasion-resistant Glove |
| Size | 11 |