Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell HyFlex™ 74-730 Off-Hand Lightweight Cut-Resistant Gloves

Catalog No. 19501021 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
11
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-501-021 10
19-501-018 7
19-501-019 8
19-501-020 9
19-501-022 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-501-021 Supplier Ansell Supplier No. 284983
This item is not returnable. View return policy

Light duty gloves offer ANSI Cut Level A4 protection and are specially designed to meet the needs associated with knife-hand and off-hand applications.

  • Manufactured using patented ZONZ™ Comfort Fit Technology
  • Dyneema™ Diamond Technology provides enhanced comfort protection in high-risk areas
  • Specially designed to meet the individual needs associated with knife-hand and off-hand applications
  • Tuff Cuff II Technology to help retain cuff shape, keeping gloves secure and reducing worker injury
  • Compliant with FDA food handling requirements
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
Construction Knit
Color Gray
Cut Resistance Rating ANSI A4
Latex Free Yes
Product Type Cut-resistant Gloves
Size 10
EN388 Standard EN 388:4X4XE, EN 388:454X
For Use With (Application) Food processing
Glove Cuff Style Tuff-Cuff™ II
Liner Material Dyneema™ Diamond Technology, Polyester, Nylon, Spandex, Stainless Steel
Model 284983
Series HyFlex™ 74-730
Length (English) 8.5 in.
Length (Metric) 21.8 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.