Learn More
Description
- Constructed of DSM Dyneema™, Spandex™ and nylon
- Ergnomic design promotes wearer comfort
- Proprietary black polyurethane coating provides sure grip
- Can be washed and reused without any loss of cut protection
Specifications
Specifications
| Coating Material | Polyurethane |
| Color | Black, White |
| Cut Resistance Rating | ANSI A2 |
| Material | Dyneema™, Lycra |
| Product Type | Cut-resistant Gloves |
| Size | 6 |
| For Use With (Application) | Glass manufacturing, assembly of white goods, Plastics injection and moulding, automotive, assembling Metal parts and components, handling of sharp edged objects, preassembling, cutting of small dry or lightly Oiled parts and maintenance |
| Glove Cuff Style | Knit |
| Hem Color | Purple |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.