Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hygromycin B from Streptomyces Hygroscopicus, MP Biomedicals™

Catalog No. MP219417091 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219417091 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219417091 Supplier MP Biomedicals, Inc Supplier No. 0219417091
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hygromycin B is an antibiotic substance isolated from Streptomyces hygroscopicus.

Its mode of action is the inhibition of protein synthesis, by inducing the misreading of the m-RNA template in the prokaryote, (E. coli, at 100 mg/mL), lower eukaryotes (e.g., yeast, at 200mg/mL) and higher eukaryotes (e.g., mammalian cells in culture). It selectively penetrates cells that have been rendered permeable by virus infection and combined with its potency to inhibit translation, it is an effective antiviral agent. Hygromycin B can be used as a dual-selection antibiotic when used in conjunction with G418 or Zeocin™ for cells transfected with two different vectors.

  • Blocks polypeptide synthesis and inhibits elongation. Specifically, it inhibits protein synthesis by interfering with translocation of the 70S ribosome and inducing misreading of the mRNA template.
  • When using this product for mammalian cell selection the working concentration is generally between 50mg/mL and 1mg/mL, but at times may go as high as 1 mg/mL.
  • The appropriate concentration for use must be determined empirically for each cell type.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31282-04-9 , 7732-18-5
Molecular Formula C20H37N3O13
Molecular Weight (g/mol) 527.524
InChI Key GRRNUXAQVGOGFE-BBMONYMYSA-N
Synonym O-6-Amino-6-deoxy-L-glycero-D-galacto-heptopyranosylidene- (1->2-3)-O-b-D- talopyranosyl- (1->5)- 2-deoxy- N3-methyl-D-streptamine
PubChem CID 134129613
IUPAC Name (3'S,3aS,4S,4'S,5'R,6R,6'R,7R,7aS)-4-[(1R,2S,3R,5S,6S)-3-amino-2,6-dihydroxy-5-(methylamino)cyclohexyl]oxy-6'-[(1S)-1-amino-2-hydroxyethyl]-6-(hydroxymethyl)spiro[4,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran-2,2'-oxane]-3',4',5',7-tetrol
SMILES CNC1CC(C(C(C1O)OC2C3C(C(C(O2)CO)O)OC4(O3)C(C(C(C(O4)C(CO)N)O)O)O)O)N

Specifications

Type Hygromycin B
Physical Form Powder
Melting Point 160°C to 180°C (literature)
Color Light Yellow
Quantity 1 g
Solubility Information Freely soluble in water (50 mg/mL), methanol,buffer solution and ethanol. Practically insoluble in less polar solvents.
Formula Weight 527.52g/mol
Percent Purity ∼95%
Grade Not Available
Recommended Storage Store at +4°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.