Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hygromycin B 80.0+%, TCI America™
SDP

Catalog No. H15091G
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 31282-04-9
Molecular Formula C20H37N3O13
Molecular Weight (g/mol) 527.524
MDL Number MFCD06795479
InChI Key GRRNUXAQVGOGFE-BBMONYMYSA-N
Synonym hygromycin b
PubChem CID 134129613
IUPAC Name (3'S,3aS,4S,4'S,5'R,6R,6'R,7R,7aS)-4-[(1R,2S,3R,5S,6S)-3-amino-2,6-dihydroxy-5-(methylamino)cyclohexyl]oxy-6'-[(1S)-1-amino-2-hydroxyethyl]-6-(hydroxymethyl)spiro[4,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran-2,2'-oxane]-3',4',5',7-tetrol
SMILES CNC1CC(C(C(C1O)OC2C3C(C(C(O2)CO)O)OC4(O3)C(C(C(C(O4)C(CO)N)O)O)O)O)N

Specifications

Melting Point 180°C
Quantity 1 g
UN Number 3462
Formula Weight 527.52
Percent Purity ≥80.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Hygromycin B
RTECSNumber WK2130000
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.