Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Hygromycin B
GREENER_CHOICESDP

Catalog No. 413750 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-375-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 41-375-0 Supplier R&D Systems™ Supplier No. 4137/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Aminoglycoside antibiotic; active against prokaryotic and eukaryotic cells

Hygromycin B is an aminoglycoside antibiotic. Active against both prokaryotic and eukaryotic cells. Inhibits protein synthesis by binding to the 30S subunit and inhibiting the ribosomal translocation step of elongation.

Chemical Identifiers

CAS 31282-04-9
Molecular Formula C20H37N3O13

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 527.52
Product Type Hygromycin B
Purity >90%
Molecular Weight (g/mol) 527.524
Chemical Name or Material Hygromycin B
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.