Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HYI Monoclonal Antibody (2A2)
GREENER_CHOICE

Catalog No. PIMA524554
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524554 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524554 Supplier Invitrogen™ Supplier No. MA524554
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MGLGAVPGRQ AAFREGLEQA VRYAKALGCP RIHLMAGRVP QGADRIAVKA EMEAVFLENL RHAAGVLAQE DLVGLLEPIN TRITDPQYFL DTPQQAAAIL QKVGRPNLQL QMDIFHWQIM DGNLTGNIRE FLPIVGHVQV AQVPGRGEPS SPGELNFPYL FQLLENEGYK GFVGCEYQPR GDTVEGLSWL RSYWDRRATQ RLASEGPHTT HVPPDSE.

Catalyzes the reversible isomerization between hydroxypyruvate and 2-hydroxy-3-oxopropanoate (also termed tartronate semialdehyde).
TRUSTED_SUSTAINABILITY

Specifications

Antigen HYI
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2A2
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Hyi
Gene Accession No. Q5T013
Gene Alias 2700033B16Rik; 6430559E15Rik; endothelial cell apoptosis protein E-CE1; HT036; hydroxypyruvate isomerase; hydroxypyruvate isomerase (putative); hydroxypyruvate isomerase homolog; hydroxypyruvate isomerase homolog (E. coli); HYI; Putative hydroxypyruvate isomerase; RGD1561416; SB156
Gene Symbols Hyi
Host Species Mouse
Immunogen HYI (AAH06140, 1 a.a. ~ 217 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81888
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.