Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HYI Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559065
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559065 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559065 Supplier Invitrogen™ Supplier No. PA559065
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALGCPRIHLM AGRVPQGADR IAVKAEMEAV FLENLRHAAG VLAQEDLVGL LEPINTRITD PQYFLDTPQQ AAAILQKVGR PNLQLQMDIF HWQIMDGNLT GNIREFLPIV GHVQVAQVPG RGEPSSPGEL NFPYLFQLLE DEGYKGFVGC Highest antigen sequence identity to the following orthologs: Mouse - 21%, Rat - 91%.

Catalyzes the reversible isomerization between hydroxypyruvate and 2-hydroxy-3-oxopropanoate (also termed tartronate semialdehyde).
TRUSTED_SUSTAINABILITY

Specifications

Antigen HYI
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Hyi
Gene Accession No. Q5T013
Gene Alias 2700033B16Rik; 6430559E15Rik; endothelial cell apoptosis protein E-CE1; HT036; hydroxypyruvate isomerase; hydroxypyruvate isomerase (putative); hydroxypyruvate isomerase homolog; hydroxypyruvate isomerase homolog (E. coli); HYI; Putative hydroxypyruvate isomerase; RGD1561416; SB156
Gene Symbols Hyi
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HYI. Recombinant protein control fragment (Product #RP-89560).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81888
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.