Learn More
Invitrogen™ HYI Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144039
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: rat liver tissue, rat kidney tissue, mouse liver tissue. IHC: human melanomar tissue, human rectal cancer tissue. ICC/IF: A431 cell. Flow: THP-1 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Catalyzes the reversible isomerization between hydroxypyruvate and 2-hydroxy-3-oxopropanoate (also termed tartronate semialdehyde).
Specifications
HYI | |
Polyclonal | |
Unconjugated | |
Hyi | |
2700033B16Rik; 6430559E15Rik; endothelial cell apoptosis protein E-CE1; HT036; hydroxypyruvate isomerase; hydroxypyruvate isomerase (putative); hydroxypyruvate isomerase homolog; hydroxypyruvate isomerase homolog (E. coli); HYI; Putative hydroxypyruvate isomerase; RGD1561416; SB156 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
500536, 68180, 81888 | |
-20°C | |
Lyophilized |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 4mg trehalose and 0.01mg sodium azide | |
Q5T013, Q8R1F5 | |
Hyi | |
E.coli-derived human HYI recombinant protein (Position: E46-Q277). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.