Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HYKK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559259
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559259 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559259 Supplier Invitrogen™ Supplier No. PA559259
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALSKPTFSEE QASALVESVF GLKVSKVRPL PSYDDQNFHV YVSKTKDGPT EYVLKISNTK ASKNPDLIEV QNHIIMFLKA Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 73%.

Catalyzes the GTP-dependent phosphorylation of 5-hydroxy-L-lysine. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen HYKK
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HYKK
Gene Accession No. A2RU49
Gene Alias 5-hydroxy-L-lysine kinase; 5-hydroxylysine kinase; AGPHD1; aminoglycoside phosphotransferase domain containing 1; aminoglycoside phosphotransferase domain-containing protein 1; C630028N24Rik; Hydroxylysine kinase; hydroxylysine kinase 1; HYKK; RGD1308677
Gene Symbols HYKK
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HYKK. Recombinant protein control fragment (Product #RP-98178).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123688
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.