Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HYLS1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
| Antigen | HYLS1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15689920
![]() |
Novus Biologicals
NBP15689920UL |
20 μL |
Each for $206.00
|
|
|||||
NBP156899
![]() |
Novus Biologicals
NBP156899 |
100 μL |
Each for $487.50
|
|
|||||
Description
HYLS1 Polyclonal specifically detects HYLS1 in Human samples. It is validated for Western Blot.Specifications
| HYLS1 | |
| Polyclonal | |
| Rabbit | |
| Q96M11 | |
| 219844 | |
| Synthetic peptides corresponding to HYLS1(hydrolethalus syndrome 1) The peptide sequence was selected from the middle region of HYLS1. Peptide sequence YFEYKRDWDSIRLPGEDHRKELRWGVREQMLCRAEPQSKPQHIYVPNNYL. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| FLJ32915HLS, hydrolethalus syndrome 1, hydrolethalus syndrome protein 1 | |
| HYLS1 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title