Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HYLS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596993
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596993 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596993 Supplier Invitrogen™ Supplier No. PA596993
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YPHVESNVPS ETVSEASQRL RKPVMKRKVL RRKPDGEVLV TDESIISESE SGTENDQDLW DLRQRLMNVQ FQEDKESSFD VSQKFNLPHE YQGISQDQLI CSLQREGMGS PAYEQDLIVA SRPKSFILPK LDQLSRNRGK TDRVARYFEY KRDWDSIRLP GEDHRKELRW GVREQMLCRA EPQSKPQHIY VPNNYLVPTE KKRSALRWGV RCDLANGVIP RKLPFPLSPS; Positive Samples: MCF-7; Cellular Location: Cytoplasm.

This gene encodes a protein localized to the cytoplasm. Mutations in this gene are associated with hydrolethalus syndrome. Multiple alternatively spliced variants, encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HYLS1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HYLS1
Gene Accession No. Q96M11
Gene Alias 3010015K02Rik; HLS; hydrolethalus syndrome 1; hydrolethalus syndrome protein 1; Hydrolethalus syndrome protein 1 homolog; Hyls1; HYLS1 centriolar and ciliogenesis associated; HYLS1, centriolar and ciliogenesis associated; hypothetical LOC538735
Gene Symbols HYLS1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 70-299 of human HYLS1 (NP_6594511).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219844
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.