Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HYLS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559480
Description
Immunogen sequence: MEELLPDGQI WANMDPEERM LAAATAFTHI CAGQGEGDVR REAQSIQYDP YSKASVAPGK RPALPVQLQY PHVESNVPSE TVSEASQRLR KPVMK Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 73%.
This gene encodes a protein localized to the cytoplasm. Mutations in this gene are associated with hydrolethalus syndrome. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifications
HYLS1 | |
Polyclonal | |
Unconjugated | |
HYLS1 | |
3010015K02Rik; HLS; hydrolethalus syndrome 1; hydrolethalus syndrome protein 1; Hydrolethalus syndrome protein 1 homolog; Hyls1; HYLS1 centriolar and ciliogenesis associated; HYLS1, centriolar and ciliogenesis associated; hypothetical LOC538735 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
219844 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.9 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96M11 | |
HYLS1 | |
Recombinant protein corresponding to Human HYLS1. Recombinant protein control fragment (Product #RP-97555). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction