Learn More
Abnova™ HYOU1 293T Cell Transient Overexpression Lysate (Denatured)
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Cell Line | 293T |
| For Use With (Application) | Western Blot |
| Description | HYOU1 293T Cell Transient Overexpression Lysate(Denatured) |
| Molecular Weight (g/mol) | Theoretical M.W. 74.69kDa |
| Plasmid | pCMV-HYOU1 full-length |
| Quality Control Testing | Transient overexpression cell lysate was tested with anti-HYOU1 antibody (Mfr. No. H00010525-B01) by Western Blot. |
| Quantity | 100 μL |
| Storage Buffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
| Host Species | Human |
| Content And Storage | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.