Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HYOU1 Monoclonal Antibody (6E3-2C3), FITC
GREENER_CHOICE

Catalog No. PIMA545339
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545339 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545339 Supplier Invitrogen™ Supplier No. MA545339
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45339 was sufficient for detection of GRP170 in 20 μg of HEK293 lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 170kDa.

HYOU1 belongs to the heat shock protein 70 family. The protein is thought to play an important role in protein folding and secretion in the ER. Since suppression of the protein is associated with accelerated apoptosis, it is also suggested to have an important cytoprotective role in hypoxia-induced cellular perturbation. This protein has been shown to be up-regulated in tumors, especially in breast tumors, and thus it is associated with tumor invasiveness. This signal peptide-lacking protein, which is only 3 amino acids shorter than the mature protein in the ER, is thought to have a housekeeping function in the cytosol. In rat, this protein localizes to both the ER by a carboxy-terminal peptide sequence and to mitochondria by an amino-terminal targeting signal.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HYOU1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 6E3-2C3
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
Gene HYOU1
Gene Accession No. Q63617, Q9JKR6, Q9Y4L1
Gene Alias 140 kDa Ca(2+)-binding protein; 150 kDa oxygen-regulated protein; 150-kDa oxygen regulated protein; 170 kDa glucose regulated protein GRP170; 170 kDa glucose-regulated protein; AI415631; Cab140; calcium binding protein 140; calcium binding protein, 140 kDa; CBP-140; DKFZp686N08236; FLJ94899; FLJ97572; Grp 170; Grp170; GRP-170; HSP 12A; HSP12A; HYOU1; hypoxia up-regulated 1; hypoxia up-regulated protein 1; Orp 150; Orp150; ORP-150; oxygen regulated protein (150kD)
Gene Symbols HYOU1
Host Species Mouse
Immunogen Recombinant Full length GRP170 Protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10525, 12282, 192235
Target Species Human, Mouse, Rat
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.