Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HYOU1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561925
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561925 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561925 Supplier Invitrogen™ Supplier No. PA561925
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLQLINDNTA TALSYGVFRR KDINTTAQNI MFYDMGSGST VCTIVTYQMV KTKEAGMQPQ LQIRGVGFDR TLGGLEMELR LRERLAGLFN EQRKGQRAKD VRENPRAMAK LLREANRLKT VLSANADHMA QIEGLMDDVD FKAKVTRV Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

HYOU1 belongs to the heat shock protein 70 family. The protein is thought to play an important role in protein folding and secretion in the ER. Since suppression of the protein is associated with accelerated apoptosis, it is also suggested to have an important cytoprotective role in hypoxia-induced cellular perturbation. This protein has been shown to be up-regulated in tumors, especially in breast tumors, and thus it is associated with tumor invasiveness. This signal peptide-lacking protein, which is only 3 amino acids shorter than the mature protein in the ER, is thought to have a housekeeping function in the cytosol. In rat, this protein localizes to both the ER by a carboxy-terminal peptide sequence and to mitochondria by an amino-terminal targeting signal.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HYOU1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HYOU1
Gene Accession No. Q9Y4L1
Gene Alias 140 kDa Ca(2+)-binding protein; 150 kDa oxygen-regulated protein; 150-kDa oxygen regulated protein; 170 kDa glucose regulated protein GRP170; 170 kDa glucose-regulated protein; AI415631; Cab140; calcium binding protein 140; calcium binding protein, 140 kDa; CBP-140; DKFZp686N08236; FLJ94899; FLJ97572; Grp 170; Grp170; GRP-170; HSP 12A; HSP12A; HYOU1; hypoxia up-regulated 1; hypoxia up-regulated protein 1; Orp 150; Orp150; ORP-150; oxygen regulated protein (150kD)
Gene Symbols HYOU1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HYOU1. Recombinant protein control fragment (Product #RP-95153).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10525
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.