Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HYOU1 Recombinant Rabbit Monoclonal Antibody (201)
GREENER_CHOICE

Catalog No. PIMA529334
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529334 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529334 Supplier Invitrogen™ Supplier No. MA529334
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human ORP150/HYOU1/HSP12A.

HYOU1 belongs to the heat shock protein 70 family. The protein is thought to play an important role in protein folding and secretion in the ER. Since suppression of the protein is associated with accelerated apoptosis, it is also suggested to have an important cytoprotective role in hypoxia-induced cellular perturbation. This protein has been shown to be up-regulated in tumors, especially in breast tumors, and thus it is associated with tumor invasiveness. This signal peptide-lacking protein, which is only 3 amino acids shorter than the mature protein in the ER, is thought to have a housekeeping function in the cytosol. In rat, this protein localizes to both the ER by a carboxy-terminal peptide sequence and to mitochondria by an amino-terminal targeting signal.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HYOU1
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 201
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene HYOU1
Gene Accession No. Q9Y4L1
Gene Alias 140 kDa Ca(2+)-binding protein; 150 kDa oxygen-regulated protein; 150-kDa oxygen regulated protein; 170 kDa glucose regulated protein GRP170; 170 kDa glucose-regulated protein; AI415631; Cab140; calcium binding protein 140; calcium binding protein, 140 kDa; CBP-140; DKFZp686N08236; FLJ94899; FLJ97572; Grp 170; Grp170; GRP-170; HSP 12A; HSP12A; HYOU1; hypoxia up-regulated 1; hypoxia up-regulated protein 1; Orp 150; Orp150; ORP-150; oxygen regulated protein (150kD)
Gene Symbols HYOU1
Host Species Rabbit
Immunogen Recombinant Human ORP150/HYOU1/HSP12A protein (Met695-Leu994).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10525
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.