Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HYPB Antibody (OTI1E1), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271850UV
Description
HYPB Monoclonal specifically detects HYPB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
HYPB | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SETD2 | |
Recombinant protein fragment corresponding to amino acids 1787-2144 of human HYPB (NP_054878) produced in E.coli. | |
0.1 mL | |
29072.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1E1 | |
Western Blot, Immunohistochemistry | |
FLJ22472, FLJ23184, FLJ45883, FLJ46217, HIF1, HIF-1EC 2.1.1.43, HIP-1, histone-lysine N-methyltransferase SETD2, hSET2, huntingtin interacting protein 1, Huntingtin yeast partner B, Huntingtin-interacting protein 1, Huntingtin-interacting protein B, HYPBp231HBP, KIAA1732SET domain-containing protein 2, KMT3AHBP231, Lysine N-methyltransferase 3A, SET domain containing 2, SET2FLJ16420 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction